Orders LinkContactUsLinkComponent
Section Block, Electron Microscopy Sciences
Section Block, 50 mL
  103305-762
Section Block, Electron Microscopy Sciences
  103305-762
 :  
Section Block, Electron Microscopy Sciences
Section Block, 50 mL
 :  
$228.59
Kit of 50
Blocking solutions based on chemically modified and fragmented ultra-pure casein. Effectively reduces unwanted binding of primary antibody and conjugates use to charged surface of the slide and tissue section.
Greatly reduces non-specific binding while preserving the specific reaction, by saturating potential non-specific protein-protein interactions.
Recommended for research and diagnostic pathology, especially for retrieved sections and polyclonal antibodies.
  • Description:
    Section Block
  • Size:
    50 mL