Orders LinkContactUsLinkComponent
SEM Bulk Specimen Holder, Electron Microscopy Sciences
SEM Bulk Specimen Holder, Electron Microscopy Sciences
X-Large Bulk Specimen Holder, 40 x 52 x 13 mm
Catalog # 103305-562
Supplier #:  75951-08
SEM Bulk Specimen Holder, Electron Microscopy Sciences
Catalog # 103305-562
SEM Bulk Specimen Holder, Electron Microscopy Sciences
SEM Bulk Specimen Holder, Electron Microscopy Sciences
X-Large Bulk Specimen Holder, 40 x 52 x 13 mm
Price
$236.25
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For clamping large irregular, bulky samples to view in a scanning electron microscope.
  • Made of machined aluminum with stainless steel allen set screws
  • Pin Diameter: 3.2 mm
Delivery information: Allen wrench included.
Specifications
  • Description:
    X-Large Bulk Specimen Holder
  • Dimensions:
    40 x 52 x 13 mm