Orders LinkContactUsLinkComponent
Graticules with Cross Scales, Electron Microscopy Sciences
Graticules with Cross Scales, Electron Microscopy Sciences
Graticule NE17, Chrome Image Surface, 20.4 mm
Catalog # 103305-404
Supplier #:  67989-20.4
Graticules with Cross Scales, Electron Microscopy Sciences
Catalog # 103305-404
Graticules with Cross Scales, Electron Microscopy Sciences
Graticules with Cross Scales, Electron Microscopy Sciences
Graticule NE17, Chrome Image Surface, 20.4 mm
Price
$240.96
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The graticule is used as a a horizontal and a vertical scale and is especially useful for measuments also in in different axis.
  • Scales in mm and inches available
  • Use on variety of different shaped objects
  • Extended size range
Specifications
  • Ø:
    20.4 mm
  • Description:
    Graticule NE17, Chrome Image Surface