SEM Clip Specimen Mounts, Electron Microscopy Sciences
Product Details & Documents
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
- Available with single and multiple clips
- Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, and more
- SEM clip mounts with multiple clips can hold larger samples or multiple smaller samples
These spring-loaded clips accommodate samples of up to 2 mm thickness, samples are held securely and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time.
Specifications
- Ø:38 mm
- Description:SEM Clip, 38 mm Pin Mount, 2 clips