Orders LinkContactUsLinkComponent
Rapid-Core Cutting Mat, Electron Microscopy Sciences
Rapid-Core Cutting Mat, Electron Microscopy Sciences
Cutting Mat, Large
Catalog # 103302-530
Supplier #:  69039-90
Rapid-Core Cutting Mat, Electron Microscopy Sciences
Catalog # 103302-530
Rapid-Core Cutting Mat, Electron Microscopy Sciences
Rapid-Core Cutting Mat, Electron Microscopy Sciences
Cutting Mat, Large
Price
$43.55
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Composite cutting mats for coring samples.
  • Protects tips and working surface
  • Reversible, inert and self-healing
  • Sterilized with ethanol
Five layer composite cutting mat will cut cleanly through an object and into the mat. Ensures the cored sample comes out completely without burrs and bits.
Specifications
  • Description:
    Cutting Mat, Large