Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Product Details & Documents
Retrieve small, specific volume samples of powders or liquids.
- Wide handles
- Unique flat bottom shape
- Dual use as weighing boats
- Non-sterile
- Translucent
Long-handled spoons are ideal for acquiring samples from bottles and jars and helping to avoid contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy.
Specifications
- Capacity:1.25 ml
- Description:Sampling Spoon
- Length:17.8 cm (7")