Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Indicator® Bin, Akro-Mils

Indicator® Bin, Akro-Mils

15 lbs., 6.75×11.625×4", Blue/orange
Catalog # 75855-588
Supplier:   Akro-Mils
Supplier #:  36462BLUE
Indicator® Bin, Akro-Mils
Catalog # 75855-588
Supplier:  Akro-Mils
Indicator® Bin, Akro-Mils
Indicator® Bin, Akro-Mils

15 lbs., 6.75×11.625×4", Blue/orange

Price
$232.03
Log in for availability
Case of 12
Product Details & Documents

The two-toned Indicator® Bin is a single-bin organizational system featuring a colored trigger side to indicate low inventory and prevent stock-outs and assembly line interruptions.

  • Bright red or orange trigger color indicates low stock levels
  • Innovative double-hopper bin can be filled or picked from both sides
  • Fit on shelving
  • Nesting
  • Made in the USA

Innovative, low-cost alternative to high-end inventory control systems. An ideal tool for implementing a Kanban system for visual process management.

Specifications
  • Capacity:
    15 lbs.
  • Color:
    Blue/orange
  • W×D×H:
    6.75×11.625×4"