T-Sue™ MicroArray Molds Kits, Simport
Product Details & Documents
T-Sue™ Microarray Mold Kits enable tissues from many donor blocks to be arrayed on a single slide.
- Can be reused hundreds of times without losing its flexibility
- Withstands temperatures between –100 to 250°C
- View many different samples on the same slide
The array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed. They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.
T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:2 mm Diameter, Red