Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
T-Sue™ MicroArray Molds Kits, Simport

T-Sue™ MicroArray Molds Kits, Simport

36 Mold Cores, 3 mm Core Diameter
Catalog # 75853-832
Supplier:   Simport Scientific
Supplier #:  M473-36PC
T-Sue™ MicroArray Molds Kits, Simport
Catalog # 75853-832
Supplier:  Simport Scientific
T-Sue™ MicroArray Molds Kits, Simport
T-Sue™ MicroArray Molds Kits, Simport

36 Mold Cores, 3 mm Core Diameter

Price
$142.48
Log in for availability
Case of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

T-Sue™ Microarray Mold Kits enable tissues from many donor blocks to be arrayed on a single slide.

  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to 250°C
  • View many different samples on the same slide

The array mold is designed to be simple and easy to use

The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed. They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.

T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.

Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.

Documents

Specifications
  • Description:
    36 Mold Cores, 3 mm Core Diameter

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.