Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2-Hexanone ≥98.0%

2-Hexanone ≥98.0%

100 mL
Catalog # TCH0114-100ML
Flammable
Health
Irritant
Supplier:   TCI America
Supplier #:  H0114-100ML
2-Hexanone ≥98.0%
Catalog # TCH0114-100ML
Supplier:  TCI America
CAS Number:  591-78-6
2-Hexanone ≥98.0%

100 mL

CAS Number:  591-78-6
Price
$105.00
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 591-78-6
MDL Number: MFCD00009482
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -57
Flash Point (°C): 23
Specific Gravity (20/20): 0.81

Other Product Names / Synonyms: Butyl Methyl Ketone

Synonyms:  Methyl butyl ketone, Butyl methyl ketone
  • CAS Number:591-78-6
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00009482
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,06060
  • Boiling Pt:127 °C
  • Melting Pt:–57 °C
  • Density:0.81 (20 °C)
  • Flash Pt:23 °C
  • Storage Temperature:Ambient
  • EINECS:209-731-1
  • UN:1224
  • ADR:3,III

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    >98.0% (GC)

Specifications

  • Size:
    100 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.