Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
α-Cyclodextrin ≥98.0%

α-Cyclodextrin ≥98.0%

100 g
Catalog # TCC0776-100G
Supplier:   TCI America
Supplier #:  C0776-100G
α-Cyclodextrin ≥98.0%
Catalog # TCC0776-100G
Supplier:  TCI America
CAS Number:  10016-20-3
α-Cyclodextrin ≥98.0%

100 g

CAS Number:  10016-20-3
Price
$319.00
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 10016-20-3
MDL Number: MFCD00078207
Molecular Formula: C36H60O30
Molecular Weight: 972.85
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 149 deg (C=1, H2O)

Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides, Cyclodextrins
Other Product Names / Synonyms: Cyclohexaamylose, Schardinger alpha-Dextrin

  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • EINECS:233-007-4

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    >98.0% (HPLC)

Specifications

  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.