Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
WIPER KNIT WITH 70% IPA AND 30% DI WATER
Catalog # 75845-736
Supplier:
Berkshire
SDS
Expand image
WIPER KNIT WITH 70% IPA AND 30% DI WATER
Catalog # 75845-736
Supplier:
Berkshire
Supplier Number:
SSPCPVP.01.12
SDS
Related Information
Documentation
SDS
ghs_SSPCPVP.01.12.pdf
Documentation