ENZYME FABRICATOR IDES 8 X 100 UNITS
Catalog # MSPP-A0FR1008
Supplier: GENOVIS, INC MS
ENZYME FABRICATOR IDES 8 X 100 UNITS
Catalog # MSPP-A0FR1008
Supplier: GENOVIS, INC MS
Supplier Number: A0-FR1-008
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
FABALACTICA ENZYME 2000 UNITS
Supplier: GENOVIS, INC MS
Trypsin/Lys-C Mix, MS Grade, Promega
Supplier: Promega Corporation
Pierce™ Immobilized Pepsin (Agarose Resin), Thermo Scientific
Supplier: Invitrogen
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics