Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

COUNT-CAL™ Count Precision Size Standards, Microgenics

10 µm diameter, 6×25 ml, Bottle, single-use, 1.05 g/cm³, 3000 particles/ml ±10%, Polystyrene microspheres in water
Catalog # MSPP-CC10PK
Marketsource
 :  CC10-PK
COUNT-CAL™ Count Precision Size Standards, Microgenics
Catalog # MSPP-CC10PK
Marketsource
CAS Number:  
COUNT-CAL™ Count Precision Size Standards, Microgenics

10 µm diameter, 6×25 ml, Bottle, single-use, 1.05 g/cm³, 3000 particles/ml ±10%, Polystyrene microspheres in water

CAS Number:  
Price
$386.15
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

COUNT-CAL™ count precision size standards, are easy to use product for validating liquid particle counters.

  • Cost-effective, convenient
  • World class particle technology solutions
  • Polystyrene microspheres are suspended in an ultra clean diluent
  • Concentration is optimized for use in light obscuration particle counters and other low concentration counting applications

Minimize contamination by eliminating the need for serial dilutions and extensive sample handling

These standards are designed for regular use in liquid particle counters to verify instrument calibration before each use, assuring the reproducibility of the particle counter by permitting a continuous record of its performance using a NIST™ traceable microsphere suspension with a precise concentration.

CC10 and CC25 are ideal for USP 788 and 788 interim testing.

 : USP, Calibration and Traceability to NIST.

Specifications

Specifications

  • Concentration:
    3000 particles/ml ±10%
  • Pack type:
    Bottle, single-use
  • Description:
    10 µm diameter
  • Size:
    6×25 ml
  • Density:
    1.05 g/cm³
  • Contents:
    Polystyrene microspheres in water
  • Cat. No.:
    MSPP-CC10PK
  • Stability:
    Contains trace amount of surfactant
  • Shelf Life:
    Product stable for a minimum of six months
  • Appearance:
    White or Colorless
  • Storage Temperature:
    2…8 °C

Specifications