$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
ASSAY MOUSE IGG 96 WELL STRIP 2 PLATE
MSPP-MIGGSCE2M
:
CELLULAR TECHNOLOGY LIMITED MS
ASSAY MOUSE IGG 96 WELL STRIP 2 PLATE
MSPP-MIGGSCE2M
:
CELLULAR TECHNOLOGY LIMITED MS
:
MIGG-SCE-2M/2