INTERFERENCE CHK STANDARD 1 200.7 125ML
Catalog # 103255-634
Supplier: Inorganic Ventures
INTERFERENCE CHK STANDARD 1 200.7 125ML
Catalog # 103255-634
Supplier: Inorganic Ventures
Supplier Number: 2007ICS-1-125ML
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Premium Aluminum Foil
Supplier: VWR International
VWR® Small Latex Bulbs
Supplier: VWR International
RT Basic Series Magnetic Stirrers, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Universal Pipette Tips
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Toluene ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International