Orders LinkContactUsLinkComponent
Microplate Vortex Mixers, Eberbach
115/230 V, 50/60 Hz, 2 deep well titer plates or 4 standard plates, 7×6×6", 0.125", 120 - 2200 rpm
  75836-356
Microplate Vortex Mixers, Eberbach
  75836-356
 :  
Microplate Vortex Mixers, Eberbach
115/230 V, 50/60 Hz, 2 deep well titer plates or 4 standard plates, 7×6×6", 0.125", 120 - 2200 rpm
These vortex lab shakers are specifically built for continuous duty and designed to produce vortex mixing of aqueous solutions in standard and deep well micro-titer plates.
  • Dynamically balanced large volume vortexing/single plane balanced small volume vortexing
  • Variable Speed digital display with digital countdown timer
  • Remote control operation
  • Tissue homogenization
  • OEM builds
  • Stainless steel models available
  • Incubator high humidity/CO₂ models
This unit has a digital programmable timer with a digital tachometer which allows timed protocols or the shaker may run continuously. The shaker has a rubber feet that prevent creeping.
  • Capacity:
    2 deep well titer plates or 4 standard plates
  • Dimensions:
    7×6×6"
  • Electrical:
    115/230 V, 50/60 Hz
  • Orbit:
    0.125"
  • Speed:
    120 - 2200 rpm