CELLROLL Roller Bottle System, Argos Technologies
CELLROLL a reliable cell culture system.
- Programmable control unit
- Rotation speeds of either 1-2 rpm or 2-6 rpm
- Equipped with an alarm system
The modular design of the CELLROLL allows the user to easily scale-up production depending on the needs: one motor unit can control between one and eight bottle racks, each rack accommodating two standard roller bottles. The moisture-resistant motor unit is available with rotation speeds of either 1-2 rpm or 2-6 rpm and is steered by a programmable control unit located outside of the incubator. In case of power failure or accidental disconnection of control and motor unit an alarm system is activated. The corrosion-free and sturdy construction makes the CELLROLL a reliable cell culture system that will keep your cells always "on the move.
: CE certified.
: Includes drive unit, two decks (for 4 roller bottles), a control unit, a power supply and a control cable.
- Description:CELLROLL Complete, 0.1-2.0 rpm