STANDARD ICP-MS COMPLETE CRFD REF 125ML
Catalog # 103255-028
Supplier: Inorganic Ventures
STANDARD ICP-MS COMPLETE CRFD REF 125ML
Catalog # 103255-028
Supplier: Inorganic Ventures
Supplier Number: IV-ICPMS-71A-125
Frequently Bought Together
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Methanol for histology
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
Eppendorf® Research® Plus Single-Channel Variable Volume Pipettors
Supplier: Eppendorf
epT.I.P.S.® Reloads, Pipette Tips
Supplier: Eppendorf
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International