Orders LinkContactUsLinkComponent
Avant™ Buffer Pouches, Advansta
Avant™ Buffer Pouches, Advansta
Avant™ Buffer Pouches - TBS, 1xbox of 20 pouch
Catalog # 103254-938
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  R-01039-020
Avant™ Buffer Pouches, Advansta
Catalog # 103254-938
Supplier:  ADVANSTA INC MS
Marketsource
CAS Number:  
Avant™ Buffer Pouches, Advansta
Avant™ Buffer Pouches - TBS, 1xbox of 20 pouch
CAS Number:  
Price
$76.15
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Convenient, fast, and pure buffer preparation.
  • Molecular grade buffers of highest purity
  • Conveniently pre-measured powder in sealed pouches
  • Quickly dissolves for immediate use
  • Standardized to guarantee reproducible results for your gels and blots
Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-measured molecular biology-grade chemicals in 500 ml of deionized water to obtain a ready-to-use 1x buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.
One pouch dissolved in 500ml purified water provides 1X working buffer.
Specifications
Specifications
  • Description:
    Avant™ Buffer Pouches - TBS
  • Size:
    1xbox of 20 pouch