Protein Sample Loading Buffers, Advansta
Product Details & Documents
Pre-mixed standard Laemmli loading buffers for protein electrophoresis.
- Pre-mixed - just add equal volume to your protein sample
- Obtain reproducible results with standardized buffers
- Choose from reducing or non-reducing loading buffer
These ready-to-use Laemmli loading buffers contain Bromophenol blue to enable visualization of the dye front and monitoring of the progress of electrophoresis.
Choose from reducing or non-reducing buffers, each supplied as a 2X concentrate.
Specifications
Specifications
- Description:Reducing protein sample loading buffer (2X)
- Size:1x 1 mL