Orders LinkContactUsLinkComponent
Development Folders, Advansta
Development Folders, Advansta
Blot Development Folders, 5 x 7", 100 folders
Catalog # 103254-868
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  L-07020-100
Development Folders, Advansta
Catalog # 103254-868
Supplier:  ADVANSTA INC MS
Marketsource
Development Folders, Advansta
Blot Development Folders, 5 x 7", 100 folders
Price
$132.69
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transparent plastic supports for handling and imaging blots.
  • Get clear images by avoiding wrinkling that can happen with plastic wrap
  • Suitable for all blots, both chemiluminescence and fluorescence imaging
  • Allows safe blot handling, avoiding scratching or ripping
  • Conveniently sized to image up to 2 blots in one folder
Advansta's Blot Development Folders are transparent plastic folders that hold your blot while it is exposed to film or a CCD camera. These folders keep your blot flat for perfect film exposures, avoiding the wrinkling or bunching that can occur with plastic wrap.
The folders are not autofluorescent and are compatible with chemiluminescence and fluorescence imaging. The folders also protect blots during handling.
Specifications
  • Description:
    Blot Development Folders
  • Dimensions:
    5 x 7"
  • Size:
    100 folders