Orders LinkContactUsLinkComponent
FLASHBlot™ Transfer Buffer, Advansta
FLASHBlot™ Transfer Buffer, 1x 500 mL
  103254-864
Marketsource
 :  R-03090-D50
FLASHBlot™ Transfer Buffer, Advansta
  103254-864
Marketsource
 :  
FLASHBlot™ Transfer Buffer, Advansta
FLASHBlot™ Transfer Buffer, 1x 500 mL
 :  
$242.31
Each
Transfer buffer designed for faster, more efficient protein transfer with increased sensitivity.
  • Transfer faster than with traditional buffers - better results in 20 min
  • More sensitive detection for low abundance proteins
  • Better detection of post-translationally modified proteins
  • Better transfer of ALL proteins, including those of high molecular weight
  • Conveniently suitable for any wet transfer set-up
Compared to traditional transfer buffers, FLASHBlot Transfer Buffer provides improved transfer efficiency of all proteins, including those of high molecular weight for which is it often very difficult to achieve complete transfer.
Transfers with the FLASHBlot buffer can be performed with all traditional wet transfer apparatuses and take only 15 to 20 minutes. Using FLASHBlot, transfer is improved with both traditional wet transfer and semi-dry.
Due to the faster transfer time, FLASHBlot Transfer Buffer allows for increased protein retention on membranes resulting in more sensitive detection of low-abundance proteins.
The short transfer times - plus increased transfer efficiency and protein retention - allow for sensitive detection of posttranslationally modified proteins, which can be difficult to detect after traditional transfer protocols.
  • Description:
    FLASHBlot™ Transfer Buffer
  • Size:
    1x 500 mL