Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Protein Sample Loading Buffers, Advansta

Protein Sample Loading Buffers, Advansta

Non-reducing protein sample loading buffer (2X), 1x 5 mL
Catalog # 103254-794
Marketsource
Supplier:   ADVANSTA INC MS
Supplier #:  R-03018-B50
Protein Sample Loading Buffers, Advansta
Catalog # 103254-794
Supplier:  ADVANSTA INC MS
Marketsource
CAS Number:  
Protein Sample Loading Buffers, Advansta

Non-reducing protein sample loading buffer (2X), 1x 5 mL

CAS Number:  
Price
$73.85
Log in for availability
Each (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Pre-mixed standard Laemmli loading buffers for protein electrophoresis.

  • Pre-mixed - just add equal volume to your protein sample
  • Obtain reproducible results with standardized buffers
  • Choose from reducing or non-reducing loading buffer

These ready-to-use Laemmli loading buffers contain Bromophenol blue to enable visualization of the dye front and monitoring of the progress of electrophoresis.

Choose from reducing or non-reducing buffers, each supplied as a 2X concentrate.

Specifications

Specifications

  • Description:
    Non-reducing protein sample loading buffer (2X)
  • Size:
    1x 5 mL

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.