Mouse IgG Isotype Control
50 mg
Catalog # 103252-620
Marketsource
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PAB MOUSE IGG INVIVOMAB 95% SDS 50MG Catalog #76555-738
Product 76555-738 is a replacement for 103252-620 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications
- Size:50 mg
- Cat. No.:103252-620
- Clonality:Polyclonal
- Antibody Type:Primary
- Isotype:IgG
- Host:Mouse



