Mouse IgG Isotype Control
100 mg
Catalog # 103252-614
Marketsource
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PAB MOUSE IGG INVIVOMAB 95% SDS 100MG Catalog #76554-526
Product 76554-526 is a replacement for 103252-614 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications
- Size:100 mg
- Cat. No.:103252-614
- Clonality:Polyclonal
- Antibody Type:Primary
- Isotype:IgG
- Host:Mouse
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



