KIT ASSAY 96WELL THODXN REDUCTASE COLMIC
Catalog # 75816-706
Supplier: Cayman Chemical Company
KIT ASSAY 96WELL THODXN REDUCTASE COLMIC
Catalog # 75816-706
Supplier: Cayman Chemical Company
Supplier Number: 10007892-96
Frequently Bought Together
Static Mouse Cage, 1284L
Supplier: Tecniplast Usa
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
Reusable Filter Animal Cage Covers, Tecniplast®
Supplier: Tecniplast Usa
VWR® Shelf Liners
Supplier: VWR International
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
COVER SHOE STANDARD SZ 12-15 CS150
Supplier: ULINE MS
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
Swab, Foam Tip, Sterile
Supplier: Hardy Diagnostics
neodisher® Detergents, Miele
Supplier: Miele
KIT GLUTATHIONE REDUCTASE ASSAY 96 WELLS
Supplier: Cayman Chemical Company
KIT CUSTOM CUT 2-LAYER DRAWER LINER FOAM
Supplier: GLOBAL EQUIPMENT MS
SUTURE ABSORBABLE 3-0 / C-14 / 30IN
Supplier: COVETRUS NORTH AMERICA MS
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
FILTER PAPER BENCHKOTE 495 X 457MM PK50
Supplier: Cytiva