Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT ICEME CMPLT EXON 8 RXN INVITRO ASSAY
Catalog # 75816-082
Supplier:
Transgenomic
SDS
Expand image
KIT ICEME CMPLT EXON 8 RXN INVITRO ASSAY
Catalog # 75816-082
Supplier:
Transgenomic
Supplier Number:
716101
SDS
Related Information
Documentation
SDS
ghs_716101
Manual
ICEme Kit Quick Start Guide
Limitations-of-ICEme-Kit
Overview-of-Multiplexed-ICE-COLD-PCR-MX-ICP
Positive-Control-for-ICEme-Kit
Reagents-and-Equipment-Required-for-ICEme-Kit
More
Datasheet
MX-ICP-Coupled-to-Multiple-Downstream-Analysis-Platforms-PMWC-2016
White-Paper-Multiplexed-ICE-COLD-PCRTM
Documentation