Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Trichophyton Agar

Trichophyton Agar

Trichophyton agar #1, 20/pack, 10.5 ml, Slant, 20×125 mm, 5 packs
Catalog # 75814-998
Supplier:   Hardy Diagnostics
Supplier #:  C51
Trichophyton Agar
Catalog # 75814-998
Supplier:  Hardy Diagnostics
CAS Number:  
Trichophyton Agar
Trichophyton Agar

Trichophyton agar #1, 20/pack, 10.5 ml, Slant, 20×125 mm, 5 packs

CAS Number:  
Price
$174.00
Log in for availability
Pack of 20 (10ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the differentiation and identification of Trichophyton spp.

Specifications

Specifications

  • Pack type:
    Slant
  • Description:
    Trichophyton agar #1
  • Dimensions:
    20×125 mm
  • Size:
    20/pack
  • Minimum Order Quantity:
    5 packs
  • Volume:
    10.5 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.