ENZYME IDES PROTEASE FABRICATOR2000UNITS
Catalog # MSPP-A0FR1020
Supplier: GENOVIS, INC MS
ENZYME IDES PROTEASE FABRICATOR2000UNITS
Catalog # MSPP-A0FR1020
Supplier: GENOVIS, INC MS
Supplier Number: A0-FR1-020
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Supplier: DISTRIBUIDORA DE PRODUCTOS MX
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Conductivity Standards, NIST, RICCA
Supplier: Ricca Chemical
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International