Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SODIUM SULFITE SOLN 1M PH10 LIQ CLR 20 L
Catalog # 103194-044
Supplier:
Aqua Solutions
SDS
Expand image
SODIUM SULFITE SOLN 1M PH10 LIQ CLR 20 L
Catalog # 103194-044
Supplier:
Aqua Solutions
Supplier Number:
HOE009
SDS
Related Information
Documentation
SDS
ghs_HOE009
Documentation