$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
POTASSIUM PERMANGANATE .0125N LIQ PUP 1L
103194-024
:
Aqua Solutions
POTASSIUM PERMANGANATE .0125N LIQ PUP 1L
103194-024
:
Aqua Solutions
:
SPX288
ghs_SPX288