Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POTASSIUM PERMANGANATE .0125M DRK PUP 1L
Catalog # 103194-022
Supplier:
Aqua Solutions
SDS
Expand image
POTASSIUM PERMANGANATE .0125M DRK PUP 1L
Catalog # 103194-022
Supplier:
Aqua Solutions
Supplier Number:
SPX289
SDS
Related Information
Documentation
SDS
ghs_SPX289
Documentation