Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acetone ≥99.5%, Technical Grade

Acetone ≥99.5%, Technical Grade

Metal drum, 200 L
Catalog # 103193-992
Flammable
Irritant
Marketsource
Supplier:   Aqua Solutions
Supplier #:  A1501-200L
Acetone ≥99.5%, Technical Grade
Catalog # 103193-992
Supplier:  Aqua Solutions
Marketsource
CAS Number:  67-64-1
Acetone ≥99.5%, Technical Grade
Acetone ≥99.5%, Technical Grade

Metal drum, 200 L

CAS Number:  67-64-1
Price
$2,152.29
Log in for availability
Each (200l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • UN:1090
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Assay:
    99.5% Min.
  • Color (APHA):
    Max. 10
  • Solubility in Water:
    Passes Test
  • Aldehyde (As HCHO):
    Max. 0.002%
  • Isopropyl alcohol:
    Max. 0.05 %
  • Evaporation residue:
    Max. 0.001%
  • Titrable acid:
    Max. 0.0003 meq/g
  • Titrable base:
    Max. 0.0006 meq/g
  • Water:
    Max. 0.5 %

Specifications

  • Pack type:
    Metal drum
  • Size:
    200 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.