Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEDIUM MCNKY AGR 0.5%BILE SALT W/OCV 5KG
Catalog # 75803-720
Supplier:
HiMedia
SDS
Expand image
MEDIUM MCNKY AGR 0.5%BILE SALT W/OCV 5KG
Catalog # 75803-720
Supplier:
HiMedia
Supplier Number:
GM082A-5KG
SDS
Related Information
Documentation
SDS
ghs_GM082A.pdf
Documentation