Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEDIUM GRNLT BRAIN HEART INFUS AGAR 500G
Catalog # 75803-496
Supplier:
HiMedia
SDS
Certificates
Expand image
MEDIUM GRNLT BRAIN HEART INFUS AGAR 500G
Catalog # 75803-496
Supplier:
HiMedia
Supplier Number:
GM211-500G
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_GM211.pdf
Documentation