$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MEDIUM GRNLT KF STREP AGAR BASE 2.5KG
75803-308
:
HiMedia
MEDIUM GRNLT KF STREP AGAR BASE 2.5KG
75803-308
:
HiMedia
:
GM1007-2.5KG
ghs_GM1007.pdf