$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MEDIUM NUT BROTH PH 6.9 W/O NACL 5 KG
75803-276
:
HiMedia
MEDIUM NUT BROTH PH 6.9 W/O NACL 5 KG
75803-276
:
HiMedia
:
GM088-5KG
ghs_GM088.pdf