Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEDIUM NUT BROTH PH 6.9 W/O NACL 2.5 KG
Catalog # 75803-272
Supplier:
HiMedia
SDS
Expand image
MEDIUM NUT BROTH PH 6.9 W/O NACL 2.5 KG
Catalog # 75803-272
Supplier:
HiMedia
Supplier Number:
GM088-2.5KG
SDS
Related Information
Documentation
SDS
ghs_GM088.pdf
Documentation