Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DEODORANT AUTCLV FRESHCLAVE APPLE SNTD
Catalog # 75802-996
Supplier:
Hardy Diagnostics
SDS
Expand image
DEODORANT AUTCLV FRESHCLAVE APPLE SNTD
Catalog # 75802-996
Supplier:
Hardy Diagnostics
Supplier Number:
FC604
SDS
Related Information
Documentation
SDS
ghs_FC604
Documentation