CALIBRATOR MINDRAY SC-CAL PLUS 2X3ML
Catalog # 75802-886
Supplier: MEDTEST DX, INC.
CALIBRATOR MINDRAY SC-CAL PLUS 2X3ML
Catalog # 75802-886
Supplier: MEDTEST DX, INC.
Supplier Number: SCCAL2US
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® Trypsin
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek