Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Biox Freezer Boxes

Biox Freezer Boxes

Freezer Box, Polypropylene, 81, 5 cm (2")
Catalog # 75799-928
Supplier:   TWD Scientific
Supplier #:  BX90-9081
Biox Freezer Boxes
Catalog # 75799-928
Supplier:  TWD Scientific
Biox Freezer Boxes
Biox Freezer Boxes

Freezer Box, Polypropylene, 81, 5 cm (2")

Price
$107.80
Log in for availability
Case of 20
Product Details & Documents

Freezer Boxes are designed to provide easy inventory management of cryogenic vials used in tissue culture, pharmaceutical research, and other biotechnology labs.

  • Store in mechanical freezers (PC/PP) or liquid nitrogen (PC)
  • Vented boxes allow rapid cooling and full drainage
  • Chemically resistant to alcohols and mild solvents
  • Grid on lid for easy identification

Plastic freezer boxes are specifically designed for Liquid nitrogen (PC) and mechanical freezer (PP) storage.

Specifications
  • No. of Holes:
    81
  • Description:
    Freezer Box, Polypropylene
  • Height:
    5 cm (2")