Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Biox Freezer Boxes

Biox Freezer Boxes

Freezer Box, Polycarbonate, 81, 5 cm (2")
Catalog # 75799-922
Supplier:   TWD Scientific
Supplier #:  BX90-9009-1
Biox Freezer Boxes
Catalog # 75799-922
Supplier:  TWD Scientific
Biox Freezer Boxes
Biox Freezer Boxes

Freezer Box, Polycarbonate, 81, 5 cm (2")

Price
$245.44
Log in for availability
Case of 20
Product Details & Documents

Freezer Boxes are designed to provide easy inventory management of cryogenic vials used in tissue culture, pharmaceutical research, and other biotechnology labs.

  • Store in mechanical freezers (PC/PP) or liquid nitrogen (PC)
  • Vented boxes allow rapid cooling and full drainage
  • Chemically resistant to alcohols and mild solvents
  • Grid on lid for easy identification

Plastic freezer boxes are specifically designed for Liquid nitrogen (PC) and mechanical freezer (PP) storage.

Specifications
  • No. of Holes:
    81
  • Description:
    Freezer Box, Polycarbonate
  • Height:
    5 cm (2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.