Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Biox Transfer Pipettes

Biox Transfer Pipettes

160 mm, Non sterile, To 3 mL, Bulk
Catalog # 75799-906
Supplier:   TWD Scientific
Supplier #:  BX30-0138-1
Biox Transfer Pipettes
Catalog # 75799-906
Supplier:  TWD Scientific
Biox Transfer Pipettes
Biox Transfer Pipettes

160 mm, Non sterile, To 3 mL, Bulk

Price
$226.05
Log in for availability
Case of 2,000
Product Details & Documents

Transfer Pipettes are perfect for quick and safe liquid transfer.

  • Disposable
  • Polyethylene construction
  • Uniform drop size
  • 100% inert for accurate sampling

Polyethylene pipettes are a great alternative to pasteur pipettes or rubber bulbs.

Specifications
  • Sterility:
    Non sterile
  • Length:
    160 mm
  • Packed:
    Bulk
  • Graduations:
    To 3 mL
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.