Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN DIOL REDISEP RF GOLD 20-40U 15.5G
Catalog # 103103-322
Supplier:
Teledyne
SDS
Expand image
COLUMN DIOL REDISEP RF GOLD 20-40U 15.5G
Catalog # 103103-322
Supplier:
Teledyne
Supplier Number:
69-2203-515
SDS
Related Information
Documentation
SDS
ghs_69-2203-515
Documentation