Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Biox Cell Strainers

40 µm, Purple
  75799-940
 :  BX15-1040
Biox Cell Strainers
  75799-940
Biox Cell Strainers
Biox Cell Strainers

40 µm, Purple

 
$246.75
   
Case of 100
 

Cell Strainers feature a strong nylon mesh for optimal performance in a variety of applications.

  • Polypropyelene frame with nylon mesh
  • Gamma radiation sterilized
  • Color coded for easy pore size identification
  • Individually wrapped
  • Applications in cell culture and filtration of various cells

Perfect fit for 50 ml centrifuge tubes and color coded according to the pore size.

 

 
  • Pore size:
    40 µm
  • Color:
    Purple