Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMN AM REDISEP RF GOLD 20-40 U 15.5 G
Catalog # 103103-274
Supplier:
Teledyne
SDS
Expand image
COLUMN AM REDISEP RF GOLD 20-40 U 15.5 G
Catalog # 103103-274
Supplier:
Teledyne
Supplier Number:
69-2203-505
SDS
Related Information
Documentation
SDS
ghs_69-2203-505
Documentation