Conductive Ultra Stack Bins, Quantum Storage Systems
Product Details & Documents
Easy access to stored items with hopper front bin and label holder provides fast identification
Protect your electronic components from typical electromagnetic interference, electrostatic or induced fields with these conductive polypropylene bins. The conductive material provides a barrier which these fileds cannot penetrate and prevents the build-up of electrostatic charge. The surface resistivity is less than 105 Ohms-per square.
Reinforced side walls and wide legde allow high stacking on benches or shelving. Bins are permantly conductive and are unaffected by washing with normal cleaners. Be sure all units are properly grounded accoring to your use. Black color.
Ordering information: Conductive Hand and Stack Bins in larger sizes (10804-244, 10804-246, and 10804-248) have a minimum order quantity of 4.
Specifications
- Description:Conductive Hang and Stack Bins
- Interior dimensions:4 3/8 x 3 7/16 x 2 13/16"
- Ext. dimensions:5 x 4 1/8 x 3"
Accessories
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-564QUS255CO binsLog in for availabilityPack of 1 - $162.34
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-558QUS239CO binsLog in for availabilityPack of 1 - $106.70
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-566QUS260CO binsLog in for availabilityPack of 1 - $195.57
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-562QUS250CO binsLog in for availabilityPack of 1 - $129.30
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-556QUS235CO binsLog in for availabilityPack of 1 - $65.01
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-560QUS240CO binsLog in for availabilityPack of 1 - $129.30
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-252QUS210CO binsLog in for availabilityPack of 1 - $33.41
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-256QUS230CO binsLog in for availabilityPack of 1 - $65.01
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-254QUS220CO binsLog in for availabilityPack of 1 - $36.39
Product Information
- Catalog #FitsAvailabilityPrice
- Conductive Divider for Hang and Stack Bins, BlackConductive Divider for Hang and Stack Bins, Black10804-568QUS270CO binsLog in for availabilityPack of 1 - $201.54