Orders LinkContactUsLinkComponent
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Hang and Stack Bins, 14 x 14 3/4 x 6 3/4", 14 3/4 x 16 1/2 x 7"
  10804-243
 :  QUS250CO
Conductive Ultra Stack Bins, Quantum Storage Systems
  10804-243
 :  
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Hang and Stack Bins, 14 x 14 3/4 x 6 3/4", 14 3/4 x 16 1/2 x 7"
Easy access to stored items with hopper front bin and label holder provides fast identification
Protect your electronic components from typical electromagnetic interference, electrostatic or induced fields with these conductive polypropylene bins. The conductive material provides a barrier which these fileds cannot penetrate and prevents the build-up of electrostatic charge. The surface resistivity is less than 105 Ohms-per square.
Reinforced side walls and wide legde allow high stacking on benches or shelving. Bins are permantly conductive and are unaffected by washing with normal cleaners. Be sure all units are properly grounded accoring to your use. Black color.
 : Conductive Hand and Stack Bins in larger sizes (10804-244, 10804-246, and 10804-248) have a minimum order quantity of 4.
  • Description:
    Conductive Hang and Stack Bins
  • Interior dimensions:
    14 x 14 3/4 x 6 3/4"
  • Ext. dimensions:
    14 3/4 x 16 1/2 x 7"
Accessories
  •  
    Description
    Fits
  • 10804-564
    Conductive Divider for Hang and Stack Bins, Black
    QUS255CO bins
    Pack of 1 - $162.34
  •  
    Description
    Fits
  • 10804-558
    Conductive Divider for Hang and Stack Bins, Black
    QUS239CO bins
    Pack of 1 - $106.70
  •  
    Description
    Fits
  • 10804-566
    Conductive Divider for Hang and Stack Bins, Black
    QUS260CO bins
    Pack of 1 - $195.57
  •  
    Description
    Fits
  • 10804-562
    Conductive Divider for Hang and Stack Bins, Black
    QUS250CO bins
    Pack of 1 - $129.30
  •  
    Description
    Fits
  • 10804-556
    Conductive Divider for Hang and Stack Bins, Black
    QUS235CO bins
    Pack of 1 - $65.01
  •  
    Description
    Fits
  • 10804-560
    Conductive Divider for Hang and Stack Bins, Black
    QUS240CO bins
    Pack of 1 - $129.30
  •  
    Description
    Fits
  • 10804-252
    Conductive Divider for Hang and Stack Bins, Black
    QUS210CO bins
    Pack of 1 - $33.41
  •  
    Description
    Fits
  • 10804-256
    Conductive Divider for Hang and Stack Bins, Black
    QUS230CO bins
    Pack of 1 - $65.01
  •  
    Description
    Fits
  • 10804-254
    Conductive Divider for Hang and Stack Bins, Black
    QUS220CO bins
    Pack of 1 - $36.39
  •  
    Description
    Fits
  • 10804-568
    Conductive Divider for Hang and Stack Bins, Black
    QUS270CO bins
    Pack of 1 - $201.54