Thumb Dressing Forceps, Electron Microscopy Sciences
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
- Made of premium grade, hardened stainless steel
- Feature a protective coating for extra durability and protection from rust and corrosion
- Satin-matte finish reduces eye-fatiguing glare
- Tip configuration:Serrated tips
- Overall length:305 mm